6-Bromonicotinic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
6-Bromopyridine-3-carboxylic acid, 2-Bromo-5-pyridinecarboxylic acid, 6-Bromo-3-pyridinecarboxylic acid
B2961-25G
B2961-5G
TCIAB2961-25GEA
194
EUR
TCIAB2961-25G
TCIAB2961-5G
6-Bromonicotinic acid ≥98.0% (by GC, titration analysis)
6-Bromonicotinic acid
Formula:
C₆H₄BrNO₂ Peso molecolare: 202,01 g/mol |
MDL Number:
MFCD01927100 CAS Number: 6311-35-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...