Tris(3-methylphenyl)phosphine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Tris(m-tolyl)phosphine, Tri-m-tolylphosphine
T1025-25G
T1025-5G
TCIAT1025-25GEA
126
EUR
TCIAT1025-25G
TCIAT1025-5G
Tris(3-methylphenyl)phosphine ≥98.0% (by GC)
Tris(3-methylphenyl)phosphine
Formula:
C₂₁H₂₁P Peso molecolare: 304,37 g/mol |
MDL Number:
MFCD00008532 CAS Number: 6224-63-1 EINECS: 228-312-4 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 100.0 to 103.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...