Tetraethylammonium iodide ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
TEAI, Tetrylammonium iodide
T0097-25G
T0097-500G
T0097-100G
TCIAT0097-25GEA
24
EUR
TCIAT0097-25G
TCIAT0097-500G
TCIAT0097-100G
Tetraethylammonium iodide ≥98.0% (by titrimetric analysis)
Tetraethylammonium iodide
Formula:
C₈H₂₀IN Peso molecolare: 257,16 g/mol Punto di fusione: 300 °C Densità: 1,566 g/cm³ (20 °C) |
MDL Number:
MFCD00011829 CAS Number: 68-05-3 EINECS: 200-676-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Potassium iodate Method) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...