Acetylcholine chloride ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(2-Acetoxyethyl)trimethylammonium chloride, 2-(Acetyloxy)-N,N,N-trimethylethanaminium chloride, 2-Acetoxy-N,N,N-trimethylethanaminium chloride
A0084-25G
A0084-500G
TCIAA0084-25GEA
28
EUR
TCIAA0084-25G
TCIAA0084-500G
Acetylcholine chloride ≥98.0% (by titrimetric analysis)
Acetylcholine chloride
Formula:
H₃CC(O)O(CH₂)₂N(CH₃)₃Cl Peso molecolare: 181,66 g/mol Punto di fusione: 151 °C Densità: 1,263 g/cm³ (20 °C) |
MDL Number:
MFCD00011698 CAS Number: 60-31-1 EINECS: 200-468-8 Merck Index: 14,00087 |
Risultati del test di specificazione
Appearance | White powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % (calcd.on dried substance) |
Melting point | 149.0 to 153.0 deg-C |
Solubility in Water | almost transparency |
Drying loss | max. 1.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...