Carbachol ≥99.0% (by titrimetric analysis)
Fornitore: TCI
|
Pericolo
|
Synonyms:
Carbamoylcholine chloride, Carbamylcholine chloride
Formula:
H₂NC(O)O(CH₂)₂N(CH₃)₃Cl Punto di fusione: 207 °C Densità: 1,389 g/cm³ (20 °C) |
MDL Number:
MFCD00012011 CAS Number: 51-83-2 EINECS: 200-127-3 UN: 2811 ADR: 6.1,II Merck Index: 14,01779 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 99.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...