Stampa…
Tetraethylammonium tetrafluoroborate ≥98.0% (by total nitrogen basis)
Fornitore: TCI EUROPE
Pericolo
Synonyms: Tetrylammonium tetrafluoroborate
T0983-25G T0983-5G
TCIAT0983-25GEA 50 EUR
TCIAT0983-25G TCIAT0983-5G
Tetraethylammonium tetrafluoroborate ≥98.0% (by total nitrogen basis)
Tetraethylammonium tetrafluoroborate
Formula: C₈H₂₀BF₄N
Peso molecolare: 217,06 g/mol
Punto di fusione: 365…368 °C
MDL Number: MFCD00011827
CAS Number: 429-06-1
EINECS: 207-055-1
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59e6d16dca315c620a6a158a252a38b7e4180a90&vid=59e6d16dca315c620a6a158a252a38b7e4180a90&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Solubility in Water almost transparency