Triethylsilylacetylene ≥96.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
T2387-1G
T2387-5G
TCIAT2387-1GEA
0
EUR
TCIAT2387-1G
TCIAT2387-5G
Triethylsilylacetylene ≥96.0% (by GC)
Triethylsilylacetylene
Formula:
C₈H₁₆Si Peso molecolare: 140,3 g/mol Punto di ebollizione: 136°C Flash Pt: 17°C |
MDL Number:
MFCD00075373 CAS Number: 1777-03-3 UN: 1993 ADR: 3,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...