Stampa…

Dimethyltin dichloride ≥99.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Dichlorodimethylstannane

D1338-25G D1338-500G D1338-100G
TCIAD1338-25GEA 25 EUR
TCIAD1338-25G TCIAD1338-500G TCIAD1338-100G
Dimethyltin dichloride ≥99.0% (by titrimetric analysis)
Dimethyltin dichloride
Formula: C₂H₆Cl₂Sn
Peso molecolare: 219,69 g/mol
Punto di ebollizione: 124 °C (1013 hPa)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00000517
CAS Number: 753-73-1
EINECS: 212-039-2
UN: 2928
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8d9b4e2fd523794737a5c1193a28dc972abc0a5&vid=e8d9b4e2fd523794737a5c1193a28dc972abc0a5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (Argentmetric Titration) min. 99.0 %
Melting point 107.0 to 109.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR