4-Benzyloxybenzyl alcohol ≥95.0% (by GC)
Fornitore: TCI
Synonyms:
(4-(Benzyloxy)phenyl)methanol
Formula:
C₁₄H₁₄O₂ Punto di fusione: 87…88 °C |
MDL Number:
MFCD00004654 CAS Number: 836-43-1 EINECS: 212-649-9 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 84.0 to 89.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...