4-Hydroxy-3-methoxybenzyl alcohol ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Vanillyl alcohol, 4-(Hydroxymethyl)-2-methoxyphenol, 2-Hydroxy-5-(hydroxymethyl)anisole, (4-Hydroxy-3-methoxyphenyl)methanol
V0018-25G
V0018-500G
TCIAV0018-25GEA
51
EUR
TCIAV0018-25G
TCIAV0018-500G
4-Hydroxy-3-methoxybenzyl alcohol ≥98.0% (by GC)
4-Hydroxy-3-methoxybenzyl alcohol
Formula:
C₈H₁₀O₃ Peso molecolare: 154,17 g/mol Punto di ebollizione: 293 °C (1013 hPa) Punto di fusione: 113 °C |
MDL Number:
MFCD00004659 CAS Number: 498-00-0 EINECS: 207-852-4 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Melting point | 113.0 to 116.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...