3-Methoxysalicyl alcohol ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(2-Hydroxy-3-methoxyphenyl)methanol, 2-(Hydroxymethyl)-6-methoxyphenol, 2-Hydroxy-3-methoxybenzyl alcohol, 2-Hydroxy-3-(hydroxymethyl)anisole
H1273-1G
H1273-5G
TCIAH1273-1GEA
74
EUR
TCIAH1273-1G
TCIAH1273-5G
3-Methoxysalicyl alcohol ≥98.0% (by GC)
3-Methoxysalicyl alcohol
Formula:
C₈H₁₀O₃ Peso molecolare: 154,17 g/mol Punto di ebollizione: 162°C |
MDL Number:
MFCD00008362 CAS Number: 4383-05-5 EINECS: 224-488-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 61.0 to 65.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...