Stampa…

3,5-Dinitrobenzyl alcohol ≥98.0% (by GC)

Fornitore: TCI EUROPE

D4144-25G D4144-5G
TCIAD4144-25GEA 190 EUR
TCIAD4144-25G TCIAD4144-5G
3,5-Dinitrobenzyl alcohol ≥98.0% (by GC)
3,5-Dinitrobenzyl alcohol
Formula: C₇H₆N₂O₅
Peso molecolare: 198,14 g/mol
Punto di fusione: 98 °C
MDL Number: MFCD00007235
CAS Number: 71022-43-0
EINECS: 275-131-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a2d4ed8284a26a4855131e0e965d85f1a03155c&vid=4a2d4ed8284a26a4855131e0e965d85f1a03155c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 89.0 to 93.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR