Stampa…

2-(Trifluoromethyl)benzenesulphonyl chloride ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-(Trifluoromethyl)benzene-1-sulphonyl chloride

T1892-5G T1892-1G
TCIAT1892-5GEA 78 EUR
TCIAT1892-5G TCIAT1892-1G
2-(Trifluoromethyl)benzenesulphonyl chloride ≥98.0% (by GC)
2-(Trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₄ClF₃O₂S
Peso molecolare: 244,62 g/mol
Punto di ebollizione: 126°C
MDL Number: MFCD00051696
CAS Number: 776-04-5
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=216ca4377f6fcc9f9c87164eacb267fc73ad67e4&vid=216ca4377f6fcc9f9c87164eacb267fc73ad67e4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR