4-(Trifluoromethyl)phenyl isothiocyanate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
1-Isocyanato-4-(trifluoromethyl)benzene, α,α,α-Trifluoro-p-tolyl isothiocyanate, p-Trifluoromethylphenylisothiocyanate
T2461-5G
T2461-1G
TCIAT2461-5GEA
84
EUR
TCIAT2461-5G
TCIAT2461-1G
4-(Trifluoromethyl)phenyl isothiocyanate ≥98.0% (by GC)
4-(Trifluoromethyl)phenyl isothiocyanate
Formula:
C₈H₄F₃NS Peso molecolare: 203,19 g/mol Punto di ebollizione: 207°C Punto di fusione: 29…31 °C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00039645 CAS Number: 1645-65-4 UN: 2811 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 38.0 to 44.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...