2-(Trifluoromethyl)phenyl isothiocyanate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
α,α,α-Trifluoro-o-tolyl isothiocyanate
T2459-1G
T2459-5G
TCIAT2459-1GEA
16
EUR
TCIAT2459-1G
TCIAT2459-5G
2-(Trifluoromethyl)phenyl isothiocyanate ≥98.0% (by GC)
2-(Trifluoromethyl)phenyl isothiocyanate
Formula:
C₈H₄F₃NS Peso molecolare: 203,19 g/mol Punto di ebollizione: 216°C Densità: 1,346 g/cm³ (20 °C) Flash Pt: 105°C |
MDL Number:
MFCD00039644 CAS Number: 1743-86-8 UN: 2810 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...