Stampa…

α,α,α,4-Tetrafluoro-m-toluic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

F0760-1G F0760-5G
TCIAF0760-1GEA 20 EUR
TCIAF0760-1G TCIAF0760-5G
α,α,α,4-Tetrafluoro-m-toluic acid ≥98.0% (by GC, titration analysis)
α,α,α,4-Tetrafluoro-m-toluic acid
Formula: C₈H₄F₄O₂
Peso molecolare: 208,11 g/mol
MDL Number: MFCD00061294
CAS Number: 67515-55-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24f85dcfd831dbb3aca74e4ab51536792d25900e&vid=24f85dcfd831dbb3aca74e4ab51536792d25900e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 113.0 to 117.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR