Stampa…

2'-Fluoropropiophenone ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: o-Fluoropropiophenone

F0450-25G F0450-5G
TCIAF0450-25GEA 97 EUR
TCIAF0450-25G TCIAF0450-5G
2'-Fluoropropiophenone ≥98.0% (by GC)
2'-Fluoropropiophenone
Formula: C₉H₉FO
Peso molecolare: 152,17 g/mol
Flash Pt: 79°C
MDL Number: MFCD00009893
CAS Number: 446-22-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a43219e4570625b4422e358c4048054bc38e19b3&vid=a43219e4570625b4422e358c4048054bc38e19b3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR