4-tert-Butylbenzhydrazide ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-tert-Butylbenzoic hydrazide, 4-(tert-Butyl)benzohydrazide, 4-tert-Butylbenzene-1-carbohydrazide, 4-tert-Butylbenzoic acid hydrazide, 4-tert-Butylbenzohydrazide
B2886-25G
B2886-5G
TCIAB2886-25GEA
115
EUR
TCIAB2886-25G
TCIAB2886-5G
4-tert-Butylbenzhydrazide ≥98.0% (by GC)
4-tert-Butylbenzhydrazide
Formula:
C₁₁H₁₆N₂O Peso molecolare: 192,26 g/mol |
MDL Number:
MFCD00014763 CAS Number: 43100-38-5 EINECS: 256-090-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Bromination Method) | min. 95.0 % |
Melting point | 128.0 to 132.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...