ω-Bromo-3,4-dichloroacetophenone ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-Bromo-1-(3,4-dichlorophenyl)ethan-1-one, 2-Bromo-1-(3,4-dichlorophenyl)ethanone, 2-Bromo-3',4'-dichloroacetophenone, 3,4-Dichlorophenacyl bromide, α-Bromo-3,4-dichloroacetophenone
B2837-25G
B2837-5G
TCIAB2837-25GEA
239
EUR
TCIAB2837-25G
TCIAB2837-5G
ω-Bromo-3,4-dichloroacetophenone ≥98.0% (by GC)
ω-Bromo-3,4-dichloroacetophenone
Formula:
C₈H₅BrCl₂O Peso molecolare: 267,94 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00051581 CAS Number: 2632-10-2 UN: 3261 ADR: 8,II |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Melting point | 57.0 to 62.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...