4-Nitro-o-phenylenediamine ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Nitro-1,2-phenylenediamine, 4-Nitrobenzene-1,2-diamine
D0103-25G
D0103-100G
TCIAD0103-25GEA
21
EUR
TCIAD0103-25G
TCIAD0103-100G
4-Nitro-o-phenylenediamine ≥97.0% (by GC, titration analysis)
4-Nitro-o-phenylenediamine
Formula:
C₆H₇N₃O₂ Peso molecolare: 153,14 g/mol Punto di fusione: 197…202 °C |
MDL Number:
MFCD00007724 CAS Number: 99-56-9 EINECS: 202-766-3 Merck Index: 14,06622 |
Risultati del test di specificazione
Appearance | Yellow to Amber to Dark red powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Solubility in N,N-DMF | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...