Stampa…

4-Iodobenzenesulphonyl chloride ≥93.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

Synonyms: p-Iodobenzenesulphonyl Chloride, Pipsyl chloride, 4-Iodobenzene-1-sulphonyl chloride

I0051-25G I0051-5G
TCIAI0051-25GEA 86 EUR
TCIAI0051-25G TCIAI0051-5G
4-Iodobenzenesulphonyl chloride ≥93.0% (by titrimetric analysis)
4-Iodobenzenesulphonyl chloride
Formula: C₆H₄ClIO₂S
Punto di fusione: 80…83 °C
MDL Number: MFCD00007441
CAS Number: 98-61-3
EINECS: 202-686-9
UN: 3261
ADR: 8,III
Merck Index: 14,07486
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf138280e398ca9cd00159705dbceaf4e0680673&vid=cf138280e398ca9cd00159705dbceaf4e0680673&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Argentmetric Titration) min. 93.0 %
Melting point 79.0 to 84.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR