Stampa…

4-Fluorobenzenesulphonyl chloride ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 4-Fluorobenzene-1-sulphonyl chloride

F0202-25G F0202-5G
TCIAF0202-25GEA 49 EUR
TCIAF0202-25G TCIAF0202-5G
4-Fluorobenzenesulphonyl chloride ≥97.0% (by GC, titration analysis)
4-Fluorobenzenesulphonyl chloride
Formula: C₆H₄ClFO₂S
Peso molecolare: 194,61 g/mol
Punto di ebollizione: 112°C
Punto di fusione: 36 °C
MDL Number: MFCD00007438
CAS Number: 349-88-2
EINECS: 206-493-0
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47afa8821206110b5bfc5c21f6fe89ba4e3d98b3&vid=47afa8821206110b5bfc5c21f6fe89ba4e3d98b3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %
Melting point 34.0 to 37.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR