Stampa…

4-Fluorosalicylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 4-Fluoro-2-hydroxybenzoic acid

F0637-1G F0637-25G F0637-5G
TCIAF0637-1GEA 24 EUR
TCIAF0637-1G TCIAF0637-25G TCIAF0637-5G
4-Fluorosalicylic acid ≥98.0% (by GC, titration analysis)
4-Fluorosalicylic acid
Formula: C₇H₅FO₃
MDL Number: MFCD00153168
CAS Number: 345-29-9
EINECS: 206-459-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f8f6bede8ab933c8b63b9f68ed223fc9628cbe4&vid=1f8f6bede8ab933c8b63b9f68ed223fc9628cbe4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 187.0 to 191.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR