Stampa…

Ethyl 2-fluorobenzoate ≥98.0% (by GC)

Fornitore: TCI

Synonyms: Ethyl 2-fluorobenzoate, 2-Fluorobenzoic acid ethyl ester

F0395-25G F0395-5G
TCIAF0395-25GEA 148 EUR
TCIAF0395-25G TCIAF0395-5G
Ethyl 2-fluorobenzoate ≥98.0% (by GC)
Ethyl-2-fluorobenzoate
Formula: C₉H₉FO₂
Punto di ebollizione: 221°C
MDL Number: MFCD00039214
CAS Number: 443-26-5
EINECS: 207-134-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=027f3e8b599547b2af8d4be43f3538b476386efc&vid=027f3e8b599547b2af8d4be43f3538b476386efc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR