Stampa…
2-Fluorobenzhydrazide ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 2-Fluorobenzohydrazide, 2-Fluorobenzene-1-carbohydrazide, 2-Fluorobenzoic hydrazide, 2-Fluorobenzoic acid hydrazide
F0616-25G F0616-5G
TCIAF0616-25GEA 252 EUR
TCIAF0616-25G TCIAF0616-5G
2-Fluorobenzhydrazide ≥98.0% (by GC, titration analysis)
2-Fluorobenzhydrazide
Formula: C₇H₇FN₂O
Peso molecolare: 154,14 g/mol
MDL Number: MFCD00025112
CAS Number: 446-24-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba6f6730d1f63bdc0dbd63bfba02c34a9c5fd151&vid=ba6f6730d1f63bdc0dbd63bfba02c34a9c5fd151&prid=web&clid=SAPDAM
Visualizza i codici
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 71.0 to 75.0 deg-C