Stampa…

4-(Trifluoromethoxy)benzoyl chloride ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 4-(Trifluoromethoxy)benzene-1-carbonyl chloride

T2341-1G T2341-5G
TCIAT2341-1GEA 21 EUR
TCIAT2341-1G TCIAT2341-5G
4-(Trifluoromethoxy)benzoyl chloride ≥98.0% (by GC, titration analysis)
4-(Trifluoromethoxy)benzoyl chloride
Formula: C₈H₄ClF₃O₂
Peso molecolare: 224,57 g/mol
Punto di ebollizione: 56°C
Densità: 1,42 g/cm³ (20 °C)
Flash Pt: 90°C
MDL Number: MFCD00052329
CAS Number: 36823-88-8
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1774985b43128628e93cca43592a9b6725862200&vid=1774985b43128628e93cca43592a9b6725862200&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR