Stampa…

3-(Trifluoromethoxy)benzoyl chloride ≥98.0% (by GC)

Fornitore: TCI
Pericolo

T2334-1G T2334-5G
TCIAT2334-1GEA 26 EUR
TCIAT2334-1G TCIAT2334-5G
3-(Trifluoromethoxy)benzoyl chloride ≥98.0% (by GC)
3-(Trifluoromethoxy)benzoyl chloride
Formula: C₈H₄ClF₃O₂
MDL Number: MFCD00061268
CAS Number: 86270-03-3
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e471e784ec010dd6bf0304cdefcb09a10151072c&vid=e471e784ec010dd6bf0304cdefcb09a10151072c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR