Stampa…

4-Amino-m-anisic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Amino-3-methoxybenzenecarboxylic acid

A2029-25G A2029-5G
TCIAA2029-25GEA 364 EUR
TCIAA2029-25G TCIAA2029-5G
4-Amino-m-anisic acid ≥98.0% (by HPLC, titration analysis)
4-Amino-m-anisic acid
Formula: C₈H₉NO₃
Peso molecolare: 167,16 g/mol
MDL Number: MFCD00016539
CAS Number: 2486-69-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=921d32d167572ccf12f702a075efa0f361ce2cd7&vid=921d32d167572ccf12f702a075efa0f361ce2cd7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 182.0 to 191.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR