p-Anisohydrazide ≥98.0% (by GC, titration analysis)
Fornitore: TCI
Synonyms:
4-Methoxybenzoic acid hydrazide, 4-Methoxybenzoic hydrazide, 4-Methoxybenzhydrazide, 4-Methoxybenzohydrazide, 4-Methoxybenzene-1-carbohydrazide, 4-Anisohydrazide
Formula:
C₈H₁₀N₂O₂ |
MDL Number:
MFCD00017073 CAS Number: 3290-99-1 EINECS: 221-952-5 |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 136.0 to 140.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...