Stampa…

4-Nitrobenzhydrazide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Nitrobenzoic hydrazide

N0149-25G N0149-5G
TCIAN0149-25GEA 81 EUR
TCIAN0149-25G TCIAN0149-5G
4-Nitrobenzhydrazide ≥98.0% (by HPLC, titration analysis)
4-Nitrobenzhydrazide
Formula: C₇H₇N₃O₃
Peso molecolare: 181,15 g/mol
MDL Number: MFCD00007604
CAS Number: 636-97-5
EINECS: 211-271-1

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Acetic acid almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR