Stampa…

Terephthaloyl dichloride ≥99.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

T0017-25G T0017-500G T0017-100G
TCIAT0017-25GEA 16 EUR
TCIAT0017-25G TCIAT0017-500G TCIAT0017-100G
Terephthaloyl dichloride ≥99.0% (by GC, titration analysis)
Terephthaloyl dichloride
Formula: C₈H₄Cl₂O₂
Punto di ebollizione: 266 °C (1013 hPa)
Punto di fusione: 82…83 °C
Densità: 1,34 g/cm³ (100 °C)
Flash Pt: 180 °C
MDL Number: MFCD00000693
CAS Number: 100-20-9
EINECS: 202-829-5
UN: 2923
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a16250d177acb4814841f3986e10a2bf9cfbb1f&vid=0a16250d177acb4814841f3986e10a2bf9cfbb1f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Argentmetric Titration) min. 99.0 %
Melting point 81.0 to 84.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR