Stampa…

3,5-Dichlorosalicylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,5-Dichloro-2-hydroxybenzoic acid

D1112-25G D1112-500G
TCIAD1112-25GEA 33 EUR
TCIAD1112-25G TCIAD1112-500G
3,5-Dichlorosalicylic acid ≥98.0% (by GC, titration analysis)
3,5-Dichlorosalicylic acid
Formula: C₇H₄Cl₂O₃
Peso molecolare: 207,01 g/mol
Punto di ebollizione: 319 °C (1013 hPa)
MDL Number: MFCD00002442
CAS Number: 320-72-9
EINECS: 206-281-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a0ae53487739b58f016255c682d1af6599ee83f&vid=7a0ae53487739b58f016255c682d1af6599ee83f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 225.0 to 228.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR