Stampa…

Terephthalaldehyde ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Terephthaldialdehyde, Terephthaldicarboxaldehyde, p-Phthalaldehyde

T0010-25G T0010-500G T0010-100G
TCIAT0010-25GEA 24 EUR
TCIAT0010-25G TCIAT0010-500G TCIAT0010-100G
Terephthalaldehyde ≥98.0% (by GC)
Terephthalaldehyde
Formula: C₈H₆O₂
Peso molecolare: 134,13 g/mol
Punto di ebollizione: 245 °C (1013 hPa)
Densità: 1,381 g/cm³ (20 °C)
MDL Number: MFCD00006949
CAS Number: 623-27-8
EINECS: 210-784-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0be1b1b077e795fb257d3d76a7f546d66a5bb4d4&vid=0be1b1b077e795fb257d3d76a7f546d66a5bb4d4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 113.0 to 117.0 deg-C
Solubility in hot Methanol almost transparency
Acid Value max. 3.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR