Stampa…

2-Bromobenzenesulphonyl chloride ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-Bromobenzene-1-sulphonyl chloride

B2702-25G B2702-5G
TCIAB2702-25GEA 185 EUR
TCIAB2702-25G TCIAB2702-5G
2-Bromobenzenesulphonyl chloride ≥97.0% (by GC, titration analysis)
2-Bromobenzenesulphonyl chloride
Formula: C₆H₄BrClO₂S
Peso molecolare: 255,52 g/mol
Punto di ebollizione: 117°C
Punto di fusione: 51 °C
MDL Number: MFCD00051971
CAS Number: 2905-25-1
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=850ad0c78ad90135ceb3e54b423d9615f0d54fa2&vid=850ad0c78ad90135ceb3e54b423d9615f0d54fa2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %
Melting point 49.0 to 53.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR