3-Bromophenetole ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
3-Bromophenyl ethyl ether, 1-Bromo-3-ethoxybenzene
B2062-25G
B2062-5G
TCIAB2062-25GEA
99
EUR
TCIAB2062-25G
TCIAB2062-5G
3-Bromophenetole ≥98.0% (by GC)
3-Bromophenetole
Formula:
C₈H₉BrO Peso molecolare: 201,06 g/mol Punto di ebollizione: 231°C Flash Pt: 83°C |
MDL Number:
MFCD00274287 CAS Number: 2655-84-7 EINECS: 220-186-9 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...