4-Bromo-3,5-dimethoxybenzoic acid ≥97.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
B2838-25G
B2838-5G
TCIAB2838-25GEA
50
EUR
TCIAB2838-25G
TCIAB2838-5G
4-Bromo-3,5-dimethoxybenzoic acid ≥97.0% (by HPLC, titration analysis)
4-Bromo-3,5-dimethoxybenzoic acid
Formula:
C₉H₉BrO₄ Peso molecolare: 261,07 g/mol Punto di fusione: 252°C |
MDL Number:
MFCD01632140 CAS Number: 56518-42-4 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...