Stampa…

Methyl-3-bromobenzoate ≥99.0% (by GC)

Fornitore: TCI

Synonyms: Methyl 3-bromobenzoate, 3-Bromobenzoic acid methyl ester

B0554-25G B0554-5G
TCIAB0554-25GEA 69 EUR
TCIAB0554-25G TCIAB0554-5G
Methyl-3-bromobenzoate ≥99.0% (by GC)
Methyl-3-bromobenzoate
Formula: C₈H₇BrO₂
Peso molecolare: 215.05 g/mol
Punto di fusione: 31
MDL Number: MFCD00017777
CAS Number: 618-89-3
EINECS: 210-569-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e9a914b46b24409ff451b9a5ef964edd060162&vid=c4e9a914b46b24409ff451b9a5ef964edd060162&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR