trans-2-Octen-1-ol ≥95.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Trans-2-octen-1-ol, trans-2-Octen-1-ol, trans-Oct-2-en-1-ol
O0247-10ML
O0247-100ML
TCIAO0247-10MLEA
42
EUR
TCIAO0247-10ML
TCIAO0247-100ML
trans-2-Octen-1-ol ≥95.0% (by GC)
trans-2-Octen-1-ol
Formula:
C₈H₁₆O Peso molecolare: 128,21 g/mol Punto di ebollizione: 92°C Flash Pt: 85°C |
MDL Number:
MFCD00014057 CAS Number: 18409-17-1 EINECS: 242-288-2 ADR: 3,III |
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |