Stampa…

Trifluoromethanesulphonic anhydride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Triflic anhydride

T1100-10G T1100-250G T1100-25G
TCIAT1100-10GEA 48 EUR
TCIAT1100-10G TCIAT1100-250G TCIAT1100-25G
Trifluoromethanesulphonic anhydride ≥98.0% (by titrimetric analysis)
Trifluoromethanesulphonic anhydride
Formula: C₂F₆O₅S₂
Peso molecolare: 282,14 g/mol
Punto di ebollizione: 84°C
Densità: 1,67 g/cm³ (20 °C)
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00000408
CAS Number: 358-23-6
EINECS: 206-616-8
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=916ac31ebf687ab0385917d01532e4c08db32567&vid=916ac31ebf687ab0385917d01532e4c08db32567&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR