Methyl difluoroacetate ≥97.0% (by GC)
Fornitore: TCI
|
Pericolo
|
M2021-25G
M2021-5G
TCIAM2021-25GEA
81
EUR
TCIAM2021-25G
TCIAM2021-5G
Methyl difluoroacetate ≥97.0% (by GC)
Methyl difluoroacetate
Formula:
C₃H₄F₂O₂ Peso molecolare: 110.06 g/mol Punto di ebollizione: 86°C Flash Pt: 24°C |
MDL Number:
MFCD00039276 CAS Number: 433-53-4 EINECS: 207-089-7 UN: 2924 ADR: 3 III |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...