Stampa…

Methyl laurate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester

L0015-25ML L0015-500ML
TCIAL0015-25MLEA 23 EUR
TCIAL0015-25ML TCIAL0015-500ML
Methyl laurate ≥98.0% (by GC)
Methyl laurate
Formula: C₁₃H₂₆O₂
Peso molecolare: 214,35 g/mol
Punto di ebollizione: 262°C
Punto di fusione: 5°C
Densità: 0,8694 g/cm³ (20 °C)
Flash Pt: 134°C
MDL Number: MFCD00008966
CAS Number: 111-82-0
EINECS: 203-911-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07f2130080c044337def023381daa82c6a11942a&vid=07f2130080c044337def023381daa82c6a11942a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR