Metacetamol ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
N-(3-Hydroxyphenyl)acetamide, N-Acetyl-3-aminophenol, Paracetamol, 3-Acetamidophenol, 3'-Hydroxyacetanilide
H0188-25G
H0188-500G
TCIAH0188-25GEA
17
EUR
TCIAH0188-25G
TCIAH0188-500G
Metacetamol ≥98.0% (by titrimetric analysis)
Metacetamol
Formula:
C₈H₉NO₂ Peso molecolare: 151,16 g/mol Punto di ebollizione: 351 °C (1013 hPa) Punto di fusione: 147…150 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00002263 CAS Number: 621-42-1 EINECS: 210-687-0 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 146.0 to 150.0 deg-C |
Solubility in hot Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...