4'-Methoxyacetanilide ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
N-(4-Methoxyphenyl)acetamide
A0019-25G
A0019-100G
TCIAA0019-25GEA
70
EUR
TCIAA0019-25G
TCIAA0019-100G
4'-Methoxyacetanilide ≥98.0% (by HPLC, total nitrogen)
4'-Methoxyacetanilide
Formula:
C₉H₁₁NO₂ Peso molecolare: 165,19 g/mol Punto di fusione: 137…138 °C Densità: 1,24 g/cm³ (20 °C) |
MDL Number:
MFCD00014963 CAS Number: 51-66-1 EINECS: 200-114-2 Merck Index: 14,00051 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 128.0 to 131.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...