Acetohydrazide ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
Acetic hydrazide, Acethydrazide
A0789-25G
A0789-500G
A0789-100G
TCIAA0789-25GEA
44
EUR
TCIAA0789-25G
TCIAA0789-500G
TCIAA0789-100G
Acetohydrazide ≥98.0% (by GC, titration analysis)
Acetohydrazide
Formula:
C₂H₆N₂O Peso molecolare: 74.08 g/mol Punto di ebollizione: 134°C |
MDL Number:
MFCD00007610 CAS Number: 1068-57-1 EINECS: 213-948-7 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Bromination Method) | min. 98.0 % |
Melting point | 66.0 to 70.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...