ADA ≥98.0% (by titrimetric analysis)
Fornitore: TCI
Synonyms:
2,2'-[(2-Amino-2-oxoethyl)imino]diacetic acid, N-(2-Acetamido)iminodiacetic acid, 2,2'-((2-Amino-2-oxoethyl)azanediyl)diacetic acid
Formula:
H₂NC(O)CH₂N(CH₂COOH)₂ Peso molecolare: 190.16 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00008031 CAS Number: 26239-55-4 EINECS: 247-530-0 Merck Index: 13,00148 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Absorbance | to pass test |
Solubility in hot Water | almost transparency |
Iron | max. 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...