Stampa…

Methyl chlorodifluoroacetate ≥97.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

C1586-25G C1586-5G
TCIAC1586-25GEA 51 EUR
TCIAC1586-25G TCIAC1586-5G
Methyl chlorodifluoroacetate ≥97.0% (by GC)
Methyl chlorodifluoroacetate
Formula: C₃H₃ClF₂O₂
Peso molecolare: 144,51 g/mol
Punto di ebollizione: 82°C
Densità: 1,3701 g/cm³ (18 °C)
Flash Pt: 19°C
MDL Number: MFCD00000775
CAS Number: 1514-87-0
EINECS: 216-154-9
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a04f1f34fdfd69c34101548fad4a0e8b3e5038d&vid=1a04f1f34fdfd69c34101548fad4a0e8b3e5038d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR