2'-Bromoacetanilide ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Bromoacetanilide, N-(2-Bromophenyl)acetamide
B3139-25G
B3139-5G
TCIAB3139-25GEA
248
EUR
TCIAB3139-25G
TCIAB3139-5G
2'-Bromoacetanilide ≥98.0% (by GC)
2'-Bromoacetanilide
Formula:
H₃CC(O)NH(C₆H₄Br) Peso molecolare: 214,06 g/mol |
MDL Number:
MFCD00099252 CAS Number: 614-76-6 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 100.0 to 104.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...