Stampa…

4'-Bromo-3'-chloroacetanilide ≥98.0% (by GC)

Fornitore: TCI EUROPE

B3141-25G B3141-5G
TCIAB3141-25GEA 133 EUR
TCIAB3141-25G TCIAB3141-5G
4'-Bromo-3'-chloroacetanilide ≥98.0% (by GC)
4'-Bromo-3'-chloroacetanilide
Formula: H₃CC(O)NH(C₆H₃BrCl)
MDL Number: MFCD00040848
CAS Number: 22459-81-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfc93107c66d59fd1460aeec44abecb64e953698&vid=dfc93107c66d59fd1460aeec44abecb64e953698&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 123.0 to 127.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR