Stampa…

4'-Bromo-2'-chloroacetanilide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B2413-25G B2413-5G
TCIAB2413-25GEA 120 EUR
TCIAB2413-25G TCIAB2413-5G
4'-Bromo-2'-chloroacetanilide ≥98.0% (by GC)
4'-Bromo-2'-chloroacetanilide
Formula: H₃CC(O)NH(C₆H₃BrCl)
Peso molecolare: 248,51 g/mol
Punto di fusione: 151°C
MDL Number: MFCD00040852
CAS Number: 3460-23-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR