D-(+)-Tyrosine ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
H-D-Tyr-OH
T1141-25G
T1141-5G
TCIAT1141-25GEA
111
EUR
TCIAT1141-25G
TCIAT1141-5G
D-(+)-Tyrosine ≥98.0% (by titrimetric analysis)
D-(+)-Tyrosine
Formula:
C₉H₁₁NO₃ Peso molecolare: 181.19 g/mol Punto di ebollizione: 351 Punto di fusione: 285 |
MDL Number:
MFCD00063073 CAS Number: 556-02-5 EINECS: 209-112-6 Merck Index: 14,09839 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Optical purity (LC) | min. 99.0 ee% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +10.4 to +12.0 deg (C=5, 1mol/L HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...